Merck
  • Combination antimicrobial susceptibility testing of Burkholderia cepacia complex: significance of species.
Combination antimicrobial susceptibility testing of Burkholderia cepacia complex: significance of species.
International journal of antimicrobial agents (2016-11-05)
Felicity K Abbott, Kathleen E N Milne, David A Stead, Ian M Gould
ABSTRACT
The Burkholderia cepacia complex (Bcc) is notorious for the life-threatening pulmonary infections it causes in patients with cystic fibrosis. The multidrug-resistant nature of Bcc and differing infective Bcc species make the design of appropriate treatment regimens challenging. Previous synergy studies have failed to take account of the species of Bcc isolates. Etest methodology was used to facilitate minimum inhibitory concentration (MIC) and antimicrobial combination testing on 258 isolates of Bcc, identified to species level by matrix-assisted laser desorption/ionisation time-of-flight mass spectrometry (MALDI-TOF/MS). The most active antimicrobials were trimethoprim/sulphamethoxazole, doxycycline and minocycline (52.5%, 46.4% and 45.9% of isolates susceptible, respectively). Synergy was observed in 9.2% of the 1799 combinations tested; the most common synergistic combinations were tobramycin + ceftazidime, meropenem + tobramycin and levofloxacin + piperacillin/tazobactam (35.4%, 32.3% and 22.2% synergy, respectively). Antimicrobial susceptibility analysis revealed differences between Burkholderia cenocepacia and Burkholderia multivorans. Disparity in clinical outcome during infection with these two micro-organisms necessitates further investigation into the clinical outcomes of treatment regimens in light of species identification and in vitro antimicrobial susceptibility studies.
Sigma-Aldrich
6-Mercaptohexanoic acid, 90%