Merck
  • Endo-lysosomal TRP mucolipin-1 channels trigger global ER Ca2+ release and Ca2+ influx.
Endo-lysosomal TRP mucolipin-1 channels trigger global ER Ca2+ release and Ca2+ influx.
Journal of cell science (2016-09-01)
Bethan S Kilpatrick, Elizabeth Yates, Christian Grimm, Anthony H Schapira, Sandip Patel
ABSTRACT
Transient receptor potential (TRP) mucolipins (TRPMLs), encoded by the MCOLN genes, are patho-physiologically relevant endo-lysosomal ion channels crucial for membrane trafficking. Several lines of evidence suggest that TRPMLs mediate localised Ca2+ release but their role in Ca2+ signalling is not clear. Here, we show that activation of endogenous and recombinant TRPMLs with synthetic agonists evoked global Ca2+ signals in human cells. These signals were blocked by a dominant-negative TRPML1 construct and a TRPML antagonist. We further show that, despite a predominant lysosomal localisation, TRPML1 supports both Ca2+ release and Ca2+ entry. Ca2+ release required lysosomal and ER Ca2+ stores suggesting that TRPMLs, like other endo-lysosomal Ca2+ channels, are capable of 'chatter' with ER Ca2+ channels. Our data identify new modalities for TRPML1 action.
Sigma-Aldrich
GW405833 hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film