Merck
  • The effect of PRMT1-mediated arginine methylation on the subcellular localization, stress granules, and detergent-insoluble aggregates of FUS/TLS.
The effect of PRMT1-mediated arginine methylation on the subcellular localization, stress granules, and detergent-insoluble aggregates of FUS/TLS.
PloS one (2012-11-16)
Atsushi Yamaguchi, Keiko Kitajo
ABSTRACT
Fused in sarcoma/translocated in liposarcoma (FUS/TLS) is one of causative genes for familial amyotrophic lateral sclerosis (ALS). In order to identify binding partners for FUS/TLS, we performed a yeast two-hybrid screening and found that protein arginine methyltransferase 1 (PRMT1) is one of binding partners primarily in the nucleus. In vitro and in vivo methylation assays showed that FUS/TLS could be methylated by PRMT1. The modulation of arginine methylation levels by a general methyltransferase inhibitor or conditional over-expression of PRMT1 altered slightly the nucleus-cytoplasmic ratio of FUS/TLS in cell fractionation assays. Although co-localized primarily in the nucleus in normal condition, FUS/TLS and PRMT1 were partially recruited to the cytoplasmic granules under oxidative stress, which were merged with stress granules (SGs) markers in SH-SY5Y cell. C-terminal truncated form of FUS/TLS (FUS-dC), which lacks C-terminal nuclear localization signal (NLS), formed cytoplasmic inclusions like ALS-linked FUS mutants and was partially co-localized with PRMT1. Furthermore, conditional over-expression of PRMT1 reduced the FUS-dC-mediated SGs formation and the detergent-insoluble aggregates in HEK293 cells. These findings indicate that PRMT1-mediated arginine methylation could be implicated in the nucleus-cytoplasmic shuttling of FUS/TLS and in the SGs formation and the detergent-insoluble inclusions of ALS-linked FUS/TLS mutants.
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-dimethyl-Arginine Antibody, asymmetric (ASYM24), serum, Upstate®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse