Merck
  • Rab5 activation promotes focal adhesion disassembly, migration and invasiveness in tumor cells.
Rab5 activation promotes focal adhesion disassembly, migration and invasiveness in tumor cells.
Journal of cell science (2013-07-03)
Pablo Mendoza, Rina Ortiz, Jorge Díaz, Andrew F G Quest, Lisette Leyton, Dwayne Stupack, Vicente A Torres
ABSTRACT
Migration and invasion are essential steps associated with tumor cell metastasis and increasing evidence points towards endosome trafficking being essential in this process. Indeed, the small GTPase Rab5, a crucial regulator of early endosome dynamics, promotes cell migration in vitro and in vivo. Precisely how Rab5 participates in these events remains to be determined. Considering that focal adhesions represent structures crucial to cell migration, we specifically asked whether Rab5 activation promoted focal adhesion disassembly and thereby facilitated migration and invasion of metastatic cancer cells. Pulldown and biosensor assays revealed that Rab5-GTP loading increased at the leading edge of migrating tumor cells. Additionally, targeting of Rab5 by different shRNA sequences, but not control shRNA, decreased Rab5-GTP levels, leading to reduced cell spreading, migration and invasiveness. Re-expression in knockdown cells of wild-type Rab5, but not the S34N mutant (GDP-bound), restored these properties. Importantly, Rab5 association with the focal adhesion proteins vinculin and paxillin increased during migration, and expression of wild-type, but not GDP-bound Rab5, accelerated focal adhesion disassembly, as well as FAK dephosphorylation on tyrosine 397. Finally, Rab5-driven invasiveness required focal adhesion disassembly, as treatment with the FAK inhibitor number 14 prevented Matrigel invasion and matrix metalloproteinase release. Taken together, these observations show that Rab5 activation is required to enhance cancer cell migration and invasion by promoting focal adhesion disassembly.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®