Merck
  • Embryonic liver fordin is involved in glucose glycolysis of hepatic stellate cell by regulating PI3K/Akt signaling.
Embryonic liver fordin is involved in glucose glycolysis of hepatic stellate cell by regulating PI3K/Akt signaling.
World journal of gastroenterology (2016-10-28)
Wei Tu, Jin Ye, Zhi-Jun Wang
ABSTRACT
To investigate the role of embryonic liver fordin (ELF) in liver fibrosis by regulating hepatic stellate cells (HSCs) glucose glycolysis. The expression of ELF and the glucose glycolysis-related proteins were evaluated in activated HSCs. siRNA was used to silence ELF expression in activated HSCs The expression of ELF increased remarkably in HSCs of the fibrosis mouse model and HSCs that were cultured for 3 wk ELF is involved in HSC glucose glycolysis by regulating PI3K/Akt signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human SPTBN1