Merck
  • Thrombospondin-1 mediates distal tubule hypertrophy induced by glycated albumin.
Thrombospondin-1 mediates distal tubule hypertrophy induced by glycated albumin.
The Biochemical journal (2003-12-20)
Yu-Lin Yang, Lea-Yea Chuang, Jinn-Yuh Guh, Shu-Fen Liu, Min-Yuan Hung, Tung-Nan Liao, Yu-Lun Huang
ABSTRACT
Diabetic nephropathy is characterized by early hypertrophy in both glomerular and tubuloepithelial elements. However, no studies to date have established a direct causal link between hyperglycaemia and renal hypertrophy. Our previous studies have found that high glucose does not induce cellular hypertrophy or expression of TGF-beta1 (transforming growth factor-beta1) in distal renal tubule cells [Yang, Guh, Yang, Lai, Tsai, Hung, Chang and Chuang (1998) J. Am. Soc. Nephrol. 9, 182-193]. In the present study, we used AGEs (advanced glycation end-products) to mimic long-term hyperglycaemia. Similar to glucose, AGEs did not induce TGF-beta1 mRNA in distal renal tubule cells [MDCK (Madin-Darby canine kidney) cells]; however, TGF-beta1 bioactivity was increased significantly. This result indicated post-translational regulation. Since TSP-1 (thrombospondin-1) has been demonstrated to activate latent TGF-beta1 in a variety of systems, the following experiments were performed. We found that AGEs dose-dependently increased both intracellular and extracellular levels of TSP-1. Purified TSP-1, like AGEs, increased the cellular protein content. Furthermore, anti-TSP-1 neutralizing antibodies attenuated the AGE-induced increase in TGF-beta1 bioactivity and hypertrophy. Thus TSP-1 might mediate AGE-induced distal renal tubule hypertrophy. In addition, we observed several putative transcription factor binding sites in the TSP-1 promoter, including those for AP-1 (activator protein-1), CREB (cAMP response element binding protein), NF-kappaB (nuclear factor-kappaB), SRF (serum response factor) and HSF (heat-shock factor), by sequence mapping. We used an enhancer assay to screen possible transcription factors involved. We showed that AP-1 and CREB were specifically induced by AGEs; furthermore, TFD (transcription factor decoy) for AP-1 could attenuate the AGE-induced increases in TSP-1 levels and cellular hypertrophy. Thus regulation of TSP-1 might be critical for hyperglycaemic distal tubule hypertrophy. Furthermore, TSP-1 TFD might be a potential approach to ameliorate diabetic renal hypertrophy.
Sigma-Aldrich
IgM, Lambda from murine myeloma, clone MOPC 104E, ascites fluid, lyophilized powder