Merck
  • Suppression of IGF binding protein-3 by palmitate promotes hepatic inflammatory responses.
Suppression of IGF binding protein-3 by palmitate promotes hepatic inflammatory responses.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2016-08-25)
Hae-Ki Min, Hitoshi Maruyama, Byoung Kuk Jang, Masahiko Shimada, Faridoddin Mirshahi, Shunlin Ren, Youngman Oh, Puneet Puri, Arun J Sanyal
ABSTRACT
IGF-binding protein-3 (IGFBP-3) is a liver-derived, anti-inflammatory molecule that is decreased in obesity, a key risk factor for nonalcoholic fatty liver disease (NAFLD). It was not known whether IGFBP-3 levels were altered in NAFLD, whether such alterations could be the result of lipotoxicity, and whether altered IGFBP-3 could affect pathways that are involved in hepatic and systemic inflammation. Serum IGFBP-3 was decreased in patients with NAFLD, whereas liver and circulating IL-8 levels were increased. Palmitate inhibited IGFBP-3 secretion by THP-1 macrophages and enhanced IL-8 expression. Exposure of palmitate-treated THP-1 macrophages to IGFBP-3-deficient conditioned medium led to a 20-fold increase in palmitate-induced IL-8 expression by hepatocytes. Conversely, overexpression of IGFBP-3 suppressed JNK and NF-κB activation and blocked palmitate-induced IL-8 expression in hepatocytes. Silencing IGFBP-3 in Huh7 cells enhanced JNK and NF-κB activity and increased palmitate-induced IL-8 secretion. These data indicate that IGFBP-3 serves as an anti-inflammatory brake in hepatocytes against JNK and NF-κB and limits their activation and downstream production of proinflammatory cytokines. Under lipotoxic conditions, palmitate inhibits hepatic macrophage secretion of IGFBP-3, thereby releasing the brake and enhancing palmitate-induced IL-8 synthesis and secretion.-Min, H.-K., Maruyama, H., Jang, B. K., Shimada, M., Mirshahi, F., Ren, S., Oh, Y., Puri, P., Sanyal, A. J. Suppression of IGF binding protein-3 by palmitate promotes hepatic inflammatory responses.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human IKBKB
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
SP600125, ≥98% (HPLC)