Merck
  • A critical assessment of the mechanisms governing the formation of aqueous biphasic systems composed of protic ionic liquids and polyethylene glycol.
A critical assessment of the mechanisms governing the formation of aqueous biphasic systems composed of protic ionic liquids and polyethylene glycol.
Physical chemistry chemical physics : PCCP (2016-11-03)
Ana Filipa M Cláudio, Jorge F B Pereira, Parker D McCrary, Mara G Freire, João A P Coutinho, Robin D Rogers
ABSTRACT
An extensive study on the formation of aqueous biphasic systems (ABS) using aqueous solutions of protic ionic liquids (PILs) and polyethylene glycol (PEG) was performed in order to understand the mechanisms underlying the phase separation. Aqueous solutions of PEG polymers with different molecular weights (600, 1000, 2000, and 3400 g mol
Sigma-Aldrich
Dipropylamine, 99%