Merck
  • Sulfate radical-based oxidation of fluoroquinolone antibiotics: Kinetics, mechanisms and effects of natural water matrices.
Sulfate radical-based oxidation of fluoroquinolone antibiotics: Kinetics, mechanisms and effects of natural water matrices.
Water research (2016-10-23)
Canlan Jiang, Yuefei Ji, Yuanyuan Shi, Jifei Chen, Tianming Cai
ABSTRACT
Widespread occurrence of fluoroquinolone antibiotics (FQs) in surface water, groundwater, soil and sediment has been reported and their remediation is essentially needed. Sulfate radical (SO
Sigma-Aldrich
Farnesyl pyrophosphate ammonium salt, methanol:ammonia solution, ≥95% (TLC)
Sigma-Aldrich
1-(2-Fluorophenyl)piperazine, 97%