- A Fas-associated death domain protein-dependent mechanism mediates the apoptotic action of non-steroidal anti-inflammatory drugs in the human leukemic Jurkat cell line.
A Fas-associated death domain protein-dependent mechanism mediates the apoptotic action of non-steroidal anti-inflammatory drugs in the human leukemic Jurkat cell line.
The Journal of biological chemistry (2001-08-22)
Z Han, P Pantazis, J H Wyche, N Kouttab, V J Kidd, E A Hendrickson
PMID11514566
ABSTRACT
Non-steroidal anti-inflammatory drugs (NSAIDs) are inhibitors of cyclooxygenase-1 and -2 and are useful for prevention and cure of cancers, especially colon and rectal cancers. The NSAIDs indomethacin and sulindac sulfide have been shown to induce apoptosis of colon epithelial cancer cells by a Bax-dependent mechanism that involves mitochondria-mediated activation of a caspase-9-dependent pathway. In this report, we demonstrate that indomethacin and sulindac sulfide induce apoptosis of human leukemic Jurkat cells by a mechanism that requires the Fas-associated Death Domain Protein-mediated activation of a caspase-8-dependent pathway. Therefore, NSAIDs induce apoptosis by different mechanisms depending on the cell type.