Merck
  • Nickel-catalyzed asymmetric negishi cross-couplings of secondary allylic chlorides with alkylzincs.
Nickel-catalyzed asymmetric negishi cross-couplings of secondary allylic chlorides with alkylzincs.
Journal of the American Chemical Society (2008-02-09)
Sunghee Son, Gregory C Fu
ABSTRACT
Complementing previous advances in allylation chemistry, an effective nickel/Pybox catalyst for regioselective asymmetric Negishi cross-couplings of racemic secondary allylic chlorides with readily available organozinc halides has been developed. The method has been applied in two key steps of a formal total synthesis of fluvirucinine A1.
Sigma-Aldrich
2,6-Bis((S)-4,5-dihydro-4-phenethyloxazol-2-yl)pyridine, 97%
Pricing and availability is not currently available.