Merck
  • SNX4 coordinates endosomal sorting of TfnR with dynein-mediated transport into the endocytic recycling compartment.
SNX4 coordinates endosomal sorting of TfnR with dynein-mediated transport into the endocytic recycling compartment.
Nature cell biology (2007-11-13)
Colin J Traer, Anna C Rutherford, Krysten J Palmer, Thomas Wassmer, Jacqueline Oakley, Naomi Attar, Jez G Carlton, Joachim Kremerskothen, David J Stephens, Peter J Cullen
ABSTRACT
SNX-BAR proteins are a sub-family of sorting nexins implicated in endosomal sorting. Here, we establish that through its phox homology (PX) and Bin-Amphiphysin-Rvs (BAR) domains, sorting nexin-4 (SNX4) is associated with tubular and vesicular elements of a compartment that overlaps with peripheral early endosomes and the juxtanuclear endocytic recycling compartment (ERC). Suppression of SNX4 perturbs transport between these compartments and causes lysosomal degradation of the transferrin receptor (TfnR). Through an interaction with KIBRA, a protein previously shown to bind dynein light chain 1, we establish that SNX4 associates with the minus end-directed microtubule motor dynein. Although suppression of KIBRA and dynein perturbs early endosome-to-ERC transport, TfnR sorting is maintained. We propose that by driving membrane tubulation, SNX4 coordinates iterative, geometric-based sorting of the TfnR with the long-range transport of carriers from early endosomes to the ERC. Finally, these data suggest that by associating with molecular motors, SNX-BAR proteins may coordinate sorting with carrier transport between donor and recipient membranes.