Merck
  • Electrophile-induced ether transfer: a new approach to polyketide structural units.
Electrophile-induced ether transfer: a new approach to polyketide structural units.
Organic letters (2006-11-03)
Kai Liu, Richard E Taylor, Rendy Kartika
ABSTRACT
[Structure: see text] A strategically novel approach to the formation of syn-1,3-diol mono- and diethers through electrophilic activation of homoallylic alkoxymethyl ethers has been developed. The resulting polyketide-like synthetic fragments are generated in good yield and with excellent stereocontrol. A chairlike transition state is proposed to account for the high stereoselectivity. Varying the conditions of the reaction workup results in the efficient generation of mono- and diether containing structural units common to polyketide natural products.
Sigma-Aldrich
Iodine monobromide, 98%
Pricing and availability is not currently available.