Merck
  • The Histone Demethylase FBXL10 Regulates the Proliferation of Spermatogonia and Ensures Long-Term Sustainable Spermatogenesis in Mice.
The Histone Demethylase FBXL10 Regulates the Proliferation of Spermatogonia and Ensures Long-Term Sustainable Spermatogenesis in Mice.
Biology of reproduction (2016-03-18)
Manabu Ozawa, Tsuyoshi Fukuda, Reiko Sakamoto, Hiroaki Honda, Nobuaki Yoshida
ABSTRACT
The F-box and leucine-rich repeat protein 10 (Fbxl10) gene encodes a protein that catalyzes demethylation of H3K4 and H3K36. In this study, we show the important roles of FBXL10 as a histone demethylase in sustainable sperm production using mice in which the JmjC domain of Fbxl10 was deleted (Fbxl10(DeltaJ/DeltaJ)). In histological analysis, testis sections from 10-wk-old Fbxl10(DeltaJ/DeltaJ) mice appeared normal. On the other hand, testes from 7-mo-old Fbxl10(DeltaJ/DeltaJ) mice contained a greater ratio of seminiferous tubules exhibiting degeneration of spermatogenesis. Further analysis using an in vitro spermatogonia culture system, that is, germline stem cells (GSCs), revealed that Fbxl10(DeltaJ/DeltaJ) GSCs expressed a significantly higher level of P21 and P19 mRNA, cyclin-dependent kinase inhibitors and also known as cellular senescence markers, than wild-type (WT) GSCs. Furthermore, the ratio of Fbxl10(DeltaJ/DeltaJ) GSCs in G0/G1 phase was higher and the ratios in S and G2/M phases were lower than the corresponding ratios of WT GSCs, and the doubling speed of Fbxl10(DeltaJ/DeltaJ) GSCs was significantly slower than that of WT GSCs. In addition to these in vitro results, an in vivo study indicated that recovery of spermatogenesis after a transient reduction in the number of testicular germ cells by busulfan treatment was significantly slower in Fbxl10(DeltaJ/DeltaJ) mice than in WT mice. These data suggest that Fbxl10 plays important roles in long-term sustainable spermatogenesis via regulating cell cycle.
Sigma-Aldrich
Anti-JHDM1B Antibody, from rabbit, purified by affinity chromatography
Supelco
Busulfan, analytical standard, for drug analysis