Merck
  • A new p-dimethylaminocinnamaldehyde reagent formulation for the photoluminescence detection of latent fingermarks on paper.
A new p-dimethylaminocinnamaldehyde reagent formulation for the photoluminescence detection of latent fingermarks on paper.
Forensic science international (2015-08-19)
Patrick Fritz, Wilhelm van Bronswijk, Simon W Lewis
ABSTRACT
Dimethylaminocinnamaldehyde was re-evaluated as a wet contact reagent for the treatment of latent fingermarks on porous substrates. A new formulation (consisting of 0.028 g p-dimethylaminocinnamaldehyde, 0.84 mL glacial acetic acid, 6.2 mL ethyl acetate and 0.993 L 40-60 °C petroleum spirits) provides quick, sensitive and robust luminescent ridge detail within 3 h of treatment on both plain and thermal paper. Comparisons to existing formulations indicate improved visualisation and/or a more efficient process.
Sigma-Aldrich
Indan-1,2-dione, 97%