Merck
  • Synthesis of hydrophobic cross-linked polyzwitterionic acid for simultaneous sorption of Eriochrome black T and chromium ions from binary hazardous waters.
Synthesis of hydrophobic cross-linked polyzwitterionic acid for simultaneous sorption of Eriochrome black T and chromium ions from binary hazardous waters.
Journal of colloid and interface science (2016-02-09)
Tawfik A Saleh, Auwal M Muhammad, Shaikh A Ali
ABSTRACT
Hydrophobic cross-linked polyzwitterionic acid (HCPZA) containing long chain (C18) hydrophobes and residues of a glutamic acid having unquenched nitrogen valency was synthesized. Exploiting the chelating ability of the amino acid residues to scavenge toxic metals and the hydrophobic surface to scoop up the organic contaminants, the resin HCPZA was evaluated for simultaneous removal of chromium and Eriochrome black T (EBT) from wastewaters. The structure and morphology of the polymer before and after sorption were characterized by using FTIR, TGA, EDX and SEM. The effect of various parameters such as contact time, pH and initial concentrations were investigated to arrive at optimum conditions. The adsorption of Eriochrome black T and Cr (III) on HCPZA reached equilibrium in 30 min. The mechanism of adsorption was investigated using kinetic, diffusion and isotherm models. The adsorption kinetic data were described well by the pseudo-second order model and by the Freundlich isotherm model. EDX analysis confirmed the adsorption of Cr (III) and EBT on the polymer. The hydrophobic resin exhibited a remarkable simultaneous adsorption capacity for EBT and Cr (III) and thus demonstrated its potential to be a promising adsorbent for removal of dyes and heavy metal ions from wastewaters.
Sigma-Aldrich
Diallylamine, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Allyl chloride, reagent grade, 98%
Pricing and availability is not currently available.