Merck
  • Tetraploid cells from cytokinesis failure induce aneuploidy and spontaneous transformation of mouse ovarian surface epithelial cells.
Tetraploid cells from cytokinesis failure induce aneuploidy and spontaneous transformation of mouse ovarian surface epithelial cells.
Cell cycle (Georgetown, Tex.) (2012-07-18)
Lei Lv, Tianwei Zhang, Qiyi Yi, Yun Huang, Zheng Wang, Heli Hou, Huan Zhang, Wei Zheng, Qiaomei Hao, Zongyou Guo, Howard J Cooke, Qinghua Shi
ABSTRACT
Most ovarian cancers originate from the ovarian surface epithelium and are characterized by aneuploid karyotypes. Aneuploidy, a consequence of chromosome instability, is an early event during the development of ovarian cancers. However, how aneuploid cells are evolved from normal diploid cells in ovarian cancers remains unknown. In the present study, cytogenetic analyses of a mouse syngeneic ovarian cancer model revealed that diploid mouse ovarian surface epithelial cells (MOSECs) experienced an intermediate tetraploid cell stage, before evolving to aneuploid (mainly near-tetraploid) cells. Using long-term live-cell imaging followed by fluorescence in situ hybridization (FISH), we demonstrated that tetraploid cells originally arose from cytokinesis failure of bipolar mitosis in diploid cells, and gave rise to aneuploid cells through chromosome mis-segregation during both bipolar and multipolar mitoses. Injection of the late passage aneuploid MOSECs resulted in tumor formation in C57BL/6 mice. Therefore, we reveal a pathway for the evolution of diploid to aneuploid MOSECs and elucidate a mechanism for the development of near-tetraploid ovarian cancer cells.
Sigma-Aldrich
Monoclonal Anti-Cytokeratin, pan antibody produced in mouse, ascites fluid, clone PCK-26
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture