Merck
  • In vivo activation of the p53 pathway by small-molecule antagonists of MDM2.
In vivo activation of the p53 pathway by small-molecule antagonists of MDM2.
Science (New York, N.Y.) (2004-01-06)
Lyubomir T Vassilev, Binh T Vu, Bradford Graves, Daisy Carvajal, Frank Podlaski, Zoran Filipovic, Norman Kong, Ursula Kammlott, Christine Lukacs, Christian Klein, Nader Fotouhi, Emily A Liu
ABSTRACT
MDM2 binds the p53 tumor suppressor protein with high affinity and negatively modulates its transcriptional activity and stability. Overexpression of MDM2, found in many human tumors, effectively impairs p53 function. Inhibition of MDM2-p53 interaction can stabilize p53 and may offer a novel strategy for cancer therapy. Here, we identify potent and selective small-molecule antagonists of MDM2 and confirm their mode of action through the crystal structures of complexes. These compounds bind MDM2 in the p53-binding pocket and activate the p53 pathway in cancer cells, leading to cell cycle arrest, apoptosis, and growth inhibition of human tumor xenografts in nude mice.
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
MDM2 Antagonist, Nutlin-3, Racemic, The MDM2 Antagonist, Nutlin-3, Racemic, also referenced under CAS 548472-68-0, controls the biological activity of MDM2. This small molecule/inhibitor is primarily used for Cancer applications.
Pricing and availability is not currently available.