Merck
  • Upregulation of KLHDC4 Predicts a Poor Prognosis in Human Nasopharyngeal Carcinoma.
Upregulation of KLHDC4 Predicts a Poor Prognosis in Human Nasopharyngeal Carcinoma.
PloS one (2016-04-01)
Yi-Fan Lian, Jing Yuan, Qian Cui, Qi-Sheng Feng, Miao Xu, Jin-Xin Bei, Yi-Xin Zeng, Lin Feng
ABSTRACT
Kelch proteins are implicated in the pathogenesis of many human diseases, including cancer. Nasopharyngeal carcinoma (NPC) is a rare malignancy in most countries, but prevalent in southern China and certain areas of Southeast Asia. In this study, we identified Kelch Domain Containing 4 (KLHDC4), an orphan member of the kelch repeat superfamily, as a prognosis marker for NPC. We examined the expression of KLHDC4 in 168 NPC cases by immunohistochemical staining and found a substantially higher level of KLHDC4 in NPC biopsies compared to adjacent normal nasopharyngeal mucosa. KLHDC4 expression was significantly related to the T classification (P <0.05), N classification (P <0.05) and total staging (P <0.01) in NPC, and patients with higher KLHDC4 expression had poorer overall (P <0.01) and metastasis-free survival (P <0.05) rates. Knockout (KO) of KLHDC4 via CRISPR/Cas9-mediated gene editing in NPC cell line dramatically inhibited cell proliferation, colony formation in soft agar and tumor formation in nude mice. In addition, cell migration and invasion were also impaired by KLHDC4 depletion as revealed by wound healing and Transwell assay. Mechanically, loss of KLHDC4 markedly induced spontaneous apoptosis in NPC cells, as evidenced by increased levels of cleaved caspase-3 and cleaved PARP. Consistently, KLHDC4 knockout cell-derived xenografts also showed elevated cleaved caspase-3 and PARP but reduced Ki-67 staining. In conclusion, our results suggest that KLHDC4 promotes NPC oncogenesis by suppressing cellular apoptosis. Thus, KLHDC4 may serve as a prognosis biomarker and a potential therapeutic target for NPC.
Sigma-Aldrich
Pepstatin A, microbial, Purity ≥98%
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Anti-KLHDC4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)