Merck
  • Differential effects of N-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline (EEDQ) on various 5-HT receptor binding sites in the rat brain.
Differential effects of N-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline (EEDQ) on various 5-HT receptor binding sites in the rat brain.
Neuropharmacology (1994-03-01)
H Gozlan, A M Laporte, S Thibault, L E Schechter, F Bolaños, M Hamon
ABSTRACT
The effects of N-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline (EEDQ), an alkylating agent producing irreversible blockade of various membrane bound receptors in brain, were investigated on four different types of serotonin receptors, 5-HT1A, 5-HT1B, 5-HT2A and 5-HT3, in various brain regions in the rat. In addition, the fate of central benzodiazepine- and "R"-zacopride-specific binding sites was also examined in rats treated with EEDQ. Membrane binding assays and/or quantitative autoradiography with appropriate radioligands indicated that EEDQ inactivated 5-HT1A, 5-HT1B and 5-HT2A sites, but was poorly active on 5-HT3, benzodiazepine and "R" sites. Among the receptors affected by EEDQ, hippocampal 5-HT1A sites were the most sensitive to the alkylating agent (ID50 approximately 1 mg/kg i.p.), followed by the cortical 5-HT2A (ID50 approximately 3 mg/kg i.p.) and the striatal 5-HT1B (ID50 approximately 6 mg/kg i.p.) sites. Pretreatment by selective ligands partially protected hippocampal 5-HT1A sites from irreversible inactivation by EEDQ (10 mg/kg i.p.) with the following order of efficacy: WAY 100635 > spiperone > BMY 7378 > ipsapirone. Similarly, pretreatment by spiperone (5 mg/kg i.p.) also reduced the ability of EEDQ to inactivated cortical 5-HT2A receptors. Analyses of the time-course recovery of respective binding sites after EEDQ administration showed that the turnover rate of 5-HT1A sites did not significantly differ in the dorsal raphe nucleus and in various forebrain areas (hippocampus, septum, cerebral cortex; half-life: approximately 4 days), but was lower than that of cortical 5-HT2A sites (half-life: 2.9 days).
Sigma-Aldrich
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, ≥99%