Merck
  • Chemopreventive action of non-steroidal anti-inflammatory drugs on the inflammatory pathways in colon cancer.
Chemopreventive action of non-steroidal anti-inflammatory drugs on the inflammatory pathways in colon cancer.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-02-24)
Preety Ghanghas, Shelly Jain, Chandan Rana, S N Sanyal
ABSTRACT
Non-steroidal anti-inflammatory drugs (NSAIDs) are emerging as novel chemopreventive agents against a variety of cancers owing to their capability in blocking the tumor development by cellular proliferation and by promoting apoptosis. Inflammation is principal cause of colon carcinogenesis. A missing link between inflammation and cancer could be the activation of NF-κB, which is a hallmark of inflammatory response, and is commonly detected in malignant tumors. Therefore, targeting pro-inflammatory cyclooxygenase enzymes and transcription factors will be profitable as a mechanism to inhibit tumor growth. In the present study, we have studied the role of various pro-inflammatory enzymes and transcription factors in the development of the 1,2-dimethylhydrazine dihydrochloride (DMH)-induced colorectal cancer and also observed the role of three NSAIDs, viz., Celecoxib, Etoricoxib and Diclofenac. Carcinogenic changes were observed in morphological and histopathological studies, whereas protein regulations of various biomolecules were identified by immunofluorescence analysis. Apoptotic studies was done by TUNEL assay and Hoechst/PI co-staining of the isolated colonocytes. It was found that DMH-treated animals were having an over-expression of pro-inflammatory enzymes, aberrant nuclear localization of activated cell survival transcription factor, NF-κB and suppression of anti-inflammatory transcription factor PPAR-γ, thereby suggesting a marked role of inflammation in the tumor progression. However, co-administration of NSAIDs has significantly reduced the inflammatory potential of the growing neoplasm.
Sigma-Aldrich
5,5-Dimethylhydantoin, 97%
Sigma-Aldrich
N,N′-Dimethylhydrazine dihydrochloride, 98%