Merck
  • Genome-Wide Chromatin Landscape Transitions Identify Novel Pathways in Early Commitment to Osteoblast Differentiation.
Genome-Wide Chromatin Landscape Transitions Identify Novel Pathways in Early Commitment to Osteoblast Differentiation.
PloS one (2016-02-20)
Bethtrice Thompson, Lyuba Varticovski, Songjoon Baek, Gordon L Hager
ABSTRACT
Bone continuously undergoes remodeling by a tightly regulated process that involves osteoblast differentiation from Mesenchymal Stem Cells (MSC). However, commitment of MSC to osteoblastic lineage is a poorly understood process. Chromatin organization functions as a molecular gatekeeper of DNA functions. Detection of sites that are hypersensitive to Dnase I has been used for detailed examination of changes in response to hormones and differentiation cues. To investigate the early steps in commitment of MSC to osteoblasts, we used a model human temperature-sensitive cell line, hFOB. When shifted to non-permissive temperature, these cells undergo "spontaneous" differentiation that takes several weeks, a process that is greatly accelerated by osteogenic induction media. We performed Dnase I hypersensitivity assays combined with deep sequencing to identify genome-wide potential regulatory events in cells undergoing early steps of commitment to osteoblasts. Massive reorganization of chromatin occurred within hours of differentiation. Whereas ~30% of unique DHS sites were located in the promoters, the majority was outside of the promoters, designated as enhancers. Many of them were at novel genomic sites and need to be confirmed experimentally. We developed a novel method for identification of cellular networks based solely on DHS enhancers signature correlated to gene expression. The analysis of enhancers that were unique to differentiating cells led to identification of bone developmental program encompassing 147 genes that directly or indirectly participate in osteogenesis. Identification of these pathways provided an unprecedented view of genomic regulation during early steps of differentiation and changes related to WNT, AP-1 and other pathways may have therapeutic implications.
Product Description
SAFC
Basal Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Oil Red O, certified by the BSC