Merck
  • Polyelectrolyte-coated nanocapsules containing undecylenic acid: Synthesis, biocompatibility and neuroprotective properties.
Polyelectrolyte-coated nanocapsules containing undecylenic acid: Synthesis, biocompatibility and neuroprotective properties.
Colloids and surfaces. B, Biointerfaces (2015-08-02)
Marek Piotrowski, Danuta Jantas, Krzysztof Szczepanowicz, Sylwia Łukasiewicz, Władysław Lasoń, Piotr Warszyński
ABSTRACT
The main objectives of the present study were to investigate the biocompatibility of polyelectrolyte-coated nanocapsules and to evaluate the neuroprotective action of the nanoencapsulated water-insoluble neuroprotective drug-undecylenic acid (UDA), in vitro. Core-shell nanocapsules were synthesized using nanoemulsification and the layer-by-layer (LbL) technique (by saturation method). The average size of synthesized nanocapsules was around 80 nm and the concentration was 2.5 × 10(10) particles/ml. Their zeta potential values ranged from less than -30 mV for the ones with external polyanion layers through -4 mV for the PEG-ylated layers to more than 30 mV for the polycation layers. Biocompatibility of synthesized nanocarriers was evaluated in the SH-SY5Y human neuroblastoma cell line using cell viability/toxicity assays (MTT reduction, LDH release). The results obtained showed that synthesized nanocapsules coated with PLL and PGA (also PEG-ylated) were non-toxic to SH-SY5Y cells, therefore, they were used as nanocarriers for UDA. Moreover, studies with ROD/FITC-labeled polyelectrolytes demonstrated approximately 20% cellular uptake of synthetized nanocapsules. Further studies showed that nanoencapsulated form of UDA was biocompatible and protected SH-SY5Y cells against the staurosporine-induced damage in lower concentrations than those of the same drug added directly to the culture medium. These data suggest that designed nanocapsules might serve as novel, promising delivery systems for neuroprotective agents.
Sigma-Aldrich
Penicillin-Streptomycin, Solution Stabilized, with 5,000 units penicillin and 5mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B acid chloride, suitable for fluorescence, technical
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Poly-L-glutamic acid sodium salt, mol wt 50,000-100,000
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Poly-L-lysine–FITC Labeled, mol wt 30,000-70,000
Sigma-Aldrich
Triton X-100, BioXtra