- Graphitic carbon nitride polymer as a recyclable photoredox catalyst for fluoroalkylation of arenes.
Graphitic carbon nitride polymer as a recyclable photoredox catalyst for fluoroalkylation of arenes.
Heterogeneous catalysis for trifluoromethylations and perfluoroalkylations has been performed. Through the usage of cheap, metal-free and recyclable mesoporous graphitic carbon nitride (mpg-CN) it was possible to fluoroalkylate various arenes by the reductive activation of sulfonyl chlorides with visible light. Thus, we were able to demonstrate the robustness and versatility of mpg-CN as a photoredox catalyst beyond water splitting and the activation of oxygen.