Merck
  • A tandem Heck-carbocyclization/Suzuki-coupling approach to the stereoselective syntheses of asymmetric 3,3-(diarylmethylene)indolinones.
A tandem Heck-carbocyclization/Suzuki-coupling approach to the stereoselective syntheses of asymmetric 3,3-(diarylmethylene)indolinones.
The Journal of organic chemistry (2005-04-23)
Wing S Cheung, Raymond J Patch, Mark R Player
ABSTRACT
[reaction: see text] An efficient and versatile method for stereoselective synthesis of (E)-3,3-(diarylmethylene)indolinones by a palladium-catalyzed tandem Heck-carbocyclization/Suzuki-coupling sequence is presented. Factors influencing yield and selectivity, namely catalyst, coordinating ligand, and solvent, are detailed.
Sigma-Aldrich
4-Chloro-2-iodoaniline, 98%