Merck
  • The mTORC2 component rictor contributes to cisplatin resistance in human ovarian cancer cells.
The mTORC2 component rictor contributes to cisplatin resistance in human ovarian cancer cells.
PloS one (2013-10-03)
Akechai Im-aram, Lee Farrand, Seung-Min Bae, Gwonhwa Song, Yong Sang Song, Jae Yong Han, Benjamin K Tsang
ABSTRACT
Resistance to cisplatin-based therapy is a major cause of treatment failure in human ovarian cancer. A better understanding of the mechanisms of cisplatin resistance will offer new insights for novel therapeutic strategies for this deadly disease. Akt and p53 are determinants of cisplatin sensitivity. Rictor is a component of mTOR protein kinase complex 2, which is required for Akt phosphorylation (Ser473) and full activation. However, the precise role of rictor and the relationship between rictor and p53 in cisplatin resistance remains poorly understood. Here, using sensitive wild-type p53 (OV2008 and A2780s), resistant wild-type p53 (C13* and OVCAR433), and p53 compromised (A2780cp, OCC1, and SKOV-3) ovarian cancer cells, we have demonstrated that (i) rictor is a determinant of cisplatin resistance in chemosensitive human ovarian cancer cells; (ii) cisplatin down-regulates rictor content by caspase-3 cleavage and proteasomal degradation; (iii) rictor down-regulation sensitizes chemo-resistant ovarian cancer cells to cisplatin-induced apoptosis in a p53-dependent manner; (iv) rictor suppresses cisplatin-induced apoptosis and confers resistance by activating and stabilizing Akt. These findings extend current knowledge on the molecular and cellular basis of cisplatin resistance and provide a rationale basis for rictor as a potential therapeutic target for chemoresistant ovarian cancer.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Monoclonal Anti-RICTOR antibody produced in mouse, clone 1F3, purified immunoglobulin, buffered aqueous solution