Merck
  • Reconstitution of the targeting of Rab6A to the Golgi apparatus in semi-intact HeLa cells: A role of BICD2 in stabilizing Rab6A on Golgi membranes and a concerted role of Rab6A/BICD2 interactions in Golgi-to-ER retrograde transport.
Reconstitution of the targeting of Rab6A to the Golgi apparatus in semi-intact HeLa cells: A role of BICD2 in stabilizing Rab6A on Golgi membranes and a concerted role of Rab6A/BICD2 interactions in Golgi-to-ER retrograde transport.
Biochimica et biophysica acta (2015-05-13)
Mariko Matsuto, Fumi Kano, Masayuki Murata
ABSTRACT
Rab is a small GTP-binding protein family that regulates various pathways of vesicular transport. Although more than 60 Rab proteins are targeted to specific organelles in mammalian cells, the mechanisms underlying the specificity of Rab proteins for the respective organelles remain unknown. In this study, we reconstituted the Golgi targeting of Rab6A in streptolysin O (SLO)-permeabilized HeLa cells in a cytosol-dependent manner and investigated the biochemical requirements of targeting. Golgi-targeting assays identified Bicaudal-D (BICD)2, which is reportedly involved in the dynein-mediated transport of mRNAs during oogenesis and embryogenesis in Drosophila, as a cytosolic factor for the Golgi targeting of Rab6A in SLO-permeabilized HeLa cells. Subsequent immunofluorescence analyses indicated decreased amounts of the GTP-bound active form of Rab6 in BICD2-knockdown cells. In addition, fluorescence recovery after photobleaching (FRAP) analyses revealed that overexpression of the C-terminal region of BICD2 decreased the exchange rate of GFP-Rab6A between the Golgi membrane and the cytosol. Collectively, these results indicated that BICD2 facilitates the binding of Rab6A to the Golgi by stabilizing its GTP-bound form. Moreover, several analyses of vesicular transport demonstrated that Rab6A and BICD2 play crucial roles in Golgi tubule fusion with the endoplasmic reticulum (ER) in brefeldin A (BFA)-treated cells, indicating that BICD2 is involved in coat protein I (COPI)-independent Golgi-to-ER retrograde vesicular transport.
Sigma-Aldrich
Anti-BICD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1