Merck
  • Iridium catalysis for C-H bond arylation of heteroarenes with iodoarenes.
Iridium catalysis for C-H bond arylation of heteroarenes with iodoarenes.
Angewandte Chemie (International ed. in English) (2009-04-09)
Benoît Join, Takuya Yamamoto, Kenichiro Itami
ABSTRACT
Efficient couplings using equimolar quantities of each coupling partner and multiple C-H bond arylation reactions are achieved with an Ir-based catalytic system for the C-H bond arylation of electron-rich heteroarenes with iodoarenes to construct extended pi-systems. The dramatic ligand effect on reaction efficiency leads to the discovery that Crabtree's catalyst (see scheme) is the optimal catalyst precursor.
Sigma-Aldrich
1,3-Diiodobenzene, 98%