Merck
  • In vitro reconstitution of co-translational D1 insertion reveals a role of the cpSec-Alb3 translocase and Vipp1 in photosystem II biogenesis.
In vitro reconstitution of co-translational D1 insertion reveals a role of the cpSec-Alb3 translocase and Vipp1 in photosystem II biogenesis.
The Biochemical journal (2015-03-25)
Björn Walter, Athina Hristou, Marc M Nowaczyk, Danja Schünemann
ABSTRACT
Photosystem II (PS II) is a multi-subunit complex localized in the thylakoid membrane that performs the light-dependent photosynthetic charge separation. The PS II reaction centre comprises, among others, the D1 protein. De novo synthesis and repair of PS II require efficient mechanisms for transport and insertion of plastid encoded D1 into the thylakoid membrane. To elucidate the process of D1 insertion, we used an in vitro translation system derived from pea chloroplasts to reconstitute the D1 insertion. Thereby, truncated D1 encoding psbA mRNAs lacking a stop codon were translated in the presence of thylakoid membranes and the translation was stalled by addition of chloramphenicol. The generated ribosome nascent chain complexes (RNCs) were tightly associated with the thylakoids. Subsequently, these D1 insertion intermediates were enriched from solubilized thylakoids by sucrose cushion centrifugation. Immunological analyses demonstrated the presence of the cpSec translocase, Alb3, cpFtsY, cpSRP54 and Vipp1 (vesicle-inducing protein in plastids 1) in the enriched D1 insertion intermediates. A complex formation between cpSecY, Alb3, cpFtsY and Vipp1 in thylakoid membranes was shown by gel filtration chromatography, BN (Blue Native)/SDS-PAGE and co-immunoprecipitation experiments. Furthermore, a stimulating effect of recombinant Vipp1 on the formation of a D1 insertion intermediate was observed in vitro. These results suggest a co-operative function of these proteins in D1 insertion.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
SAFC
Sodium chloride solution, 5 M