Merck
  • Sex and parasites: genomic and transcriptomic analysis of Microbotryum lychnidis-dioicae, the biotrophic and plant-castrating anther smut fungus.
Sex and parasites: genomic and transcriptomic analysis of Microbotryum lychnidis-dioicae, the biotrophic and plant-castrating anther smut fungus.
BMC genomics (2015-06-17)
Michael H Perlin, Joelle Amselem, Eric Fontanillas, Su San Toh, Zehua Chen, Jonathan Goldberg, Sebastien Duplessis, Bernard Henrissat, Sarah Young, Qiandong Zeng, Gabriela Aguileta, Elsa Petit, Helene Badouin, Jared Andrews, Dominique Razeeq, Toni Gabaldón, Hadi Quesneville, Tatiana Giraud, Michael E Hood, David J Schultz, Christina A Cuomo
ABSTRACT
The genus Microbotryum includes plant pathogenic fungi afflicting a wide variety of hosts with anther smut disease. Microbotryum lychnidis-dioicae infects Silene latifolia and replaces host pollen with fungal spores, exhibiting biotrophy and necrosis associated with altering plant development. We determined the haploid genome sequence for M. lychnidis-dioicae and analyzed whole transcriptome data from plant infections and other stages of the fungal lifecycle, revealing the inventory and expression level of genes that facilitate pathogenic growth. Compared to related fungi, an expanded number of major facilitator superfamily transporters and secretory lipases were detected; lipase gene expression was found to be altered by exposure to lipid compounds, which signaled a switch to dikaryotic, pathogenic growth. In addition, while enzymes to digest cellulose, xylan, xyloglucan, and highly substituted forms of pectin were absent, along with depletion of peroxidases and superoxide dismutases that protect the fungus from oxidative stress, the repertoire of glycosyltransferases and of enzymes that could manipulate host development has expanded. A total of 14% of the genome was categorized as repetitive sequences. Transposable elements have accumulated in mating-type chromosomal regions and were also associated across the genome with gene clusters of small secreted proteins, which may mediate host interactions. The unique absence of enzyme classes for plant cell wall degradation and maintenance of enzymes that break down components of pollen tubes and flowers provides a striking example of biotrophic host adaptation.
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
Phytol, ≥97%, FG
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Phytol, 95%, mixture of isomers
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)