Merck
  • Simultaneous plasma and genital pharmacokinetics and pharmacodynamics of atazanavir and efavirenz in HIV-infected women starting therapy.
Simultaneous plasma and genital pharmacokinetics and pharmacodynamics of atazanavir and efavirenz in HIV-infected women starting therapy.
Journal of clinical pharmacology (2015-02-17)
Michael Neely, Stan Louie, Jiaao Xu, Patricia Anthony, Kasalyn Thuvamontolrat, Nicholas Mordwinkin, Andrea Kovacs
ABSTRACT
Few studies have characterized longitudinal female plasma and genital antiretroviral pharmacokinetics and pharmacodynamics. Among 20 regimen-naive HIV-infected adult women initiating atazanavir-based therapy (n = 9) or efavirenz-based therapy (n = 11), we measured blood CD4+ T lymphocytes, and paired plasma and genital HIV RNA and atazanavir or efavirenz 2 days before starting therapy and 2, 4, 7, 10, 21, 28, 60, 120, and 180 days after. The mean (range) log10 baseline plasma viral load was 4.89 copies/mL (2.64-6.09 copies/mL), and genital was3.30 (1.60-5.00). In the atazanavir and efavirenz groups, mean (SD) days to a 50% decrease in plasma viral load was 8.2 (4.9) versus 9.3 (7.4), P = .7, and in the genital tract it was 7.3 (3.5) versus 9.3 (7.7), P = .5. The median (interquartile range) plasma:genital concentration ratio for atazanavir was 0.11 (0.001-0.46) versus 0.34 (0.05-1.30) for efavirenz, P = .5. Average plasma efavirenz or atazanavir concentrations over time did not affect virologic response. Blood CD4+ percentages increased by +2.3 (P = .06) and +3.0 (P = .003) for every 1 mg/L increase in average plasma and genital drug concentration, respectively. Plasma and genital viral pharmacodynamics were similar between the groups and independent of average concentrations, but blood CD4+ response was related in particular to genital extravascular drug concentrations.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%