Merck
  • Simple and Rapid High-Yield Synthesis and Size Sorting of Multibranched Hollow Gold Nanoparticles with Highly Tunable NIR Plasmon Resonances.
Simple and Rapid High-Yield Synthesis and Size Sorting of Multibranched Hollow Gold Nanoparticles with Highly Tunable NIR Plasmon Resonances.
Small (Weinheim an der Bergstrasse, Germany) (2015-06-13)
Adam J Blanch, Markus Döblinger, Jessica Rodríguez-Fernández
ABSTRACT
Branched gold nanoparticles with sharp tips are considered excellent candidates for sensing and field enhancement applications. Here, a rapid and simple synthesis strategy is presented that generates highly branched gold nanoparticles with hollow cores and a ca.100% yield through a simple one-pot seedless reaction at room temperature in the presence of Triton X-100. It is shown that multibranched hollow gold nanoparticles of tunable dimensions, branch density and branch length can be obtained by adjusting the concentrations of the reactants. Insights into the formation mechanism point toward an aggregative type of growth involving hollow core formation first, and branching thereafter. The pronounced near-infrared (NIR) plasmon band of the nanoparticles is due to the combined contribution from hollowness and branching, and can be tuned over a wide range (≈700-2000 nm). It is also demonstrated that the high environmental sensitivity of colloidal dispersions based on multibranched hollow gold nanoparticles can be boosted even further by separating the nanoparticles into fractions of given sizes and improved monodispersity by means of a glycerol density gradient. The possibility to obtain highly monodisperse multibranched hollow gold nanoparticles with predictable dimensions (50-300 nm) and branching and, therefore, tailored NIR plasmonic properties, highlights their potential for theranostic applications.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%