Merck
  • A Novel Loading Method for Doxycycline Liposomes for Intracellular Drug Delivery: Characterization of In Vitro and In Vivo Release Kinetics and Efficacy in a J774A.1 Cell Line Model of Mycobacterium smegmatis Infection.
A Novel Loading Method for Doxycycline Liposomes for Intracellular Drug Delivery: Characterization of In Vitro and In Vivo Release Kinetics and Efficacy in a J774A.1 Cell Line Model of Mycobacterium smegmatis Infection.
Drug metabolism and disposition: the biological fate of chemicals (2015-06-03)
Rebekah K Franklin, Sarah A Marcus, Adel M Talaat, Butch K KuKanich, Ruth Sullivan, Lisa A Krugner-Higby, Timothy D Heath
ABSTRACT
Doxycycline (doxy) is used in treating intracellular and extracellular infections. Liposomal (LE) antibiotics allow low-frequency dosing and extended efficacy compared with standard (STD) formulations. We developed a novel sulfuric acid-loading method for doxycycline liposomes (LE-doxy). We hypothesized that a single s.c. injection of LE-doxy would be detectable in serum for at least 2 weeks at concentrations equal to or better than STD-doxy and would be bactericidal in an in vitro Mycobacterium smegmatis infection of J774A.1 macrophage cells. Liposomes were encapsulated by sulfuric acid gradient loading, and release kinetics were performed in vitro and in vivo. LE-doxy made using 8.25 mg/ml doxycycline loaded for 24 hours achieved 97.77% capture in 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) and 43.87% in sphingomyelin (sphing). Rats were injected s.c. with 50 mg/kg LE-doxy or 5 mg/kg STD-doxy, and serial blood samples were collected. Pharmacokinetics were analyzed using high-performance liquid chromatography. Liver and injection site skin samples were collected at euthanasia (4 weeks postinjection). Minimal histologic tissue reactions occurred after injection of STD (nonliposomal), DPPC, or sphing-doxy. DPPC-doxy had slightly faster in vitro leakage than sphing liposomes, although both were detectable at 264 hours. The mean residence time for DPPC was the highest (111.78 hours), followed by sphing (56.00 hours) and STD (6.86 hours). DPPC and sphing-doxy were detectable at 0.2 μg/ml in serum at 336 hours postadministration. LE-doxy was not toxic to J774A.1 cells in vitro and produced inhibition of viable Mycobacterium smegmatis at 24 and 48 hours. LE-doxy will require further testing in in vivo infection models.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
SAFC
Cholesterol, Plant-Derived, SyntheChol®