Merck
  • An Oligonucleotide Transfection Vector Based on HSA and PDMAEMA Conjugation: Effect of Polymer Molecular Weight on Cell Proliferation and on Multicellular Tumor Spheroids.
An Oligonucleotide Transfection Vector Based on HSA and PDMAEMA Conjugation: Effect of Polymer Molecular Weight on Cell Proliferation and on Multicellular Tumor Spheroids.
Macromolecular bioscience (2015-03-27)
Yanyan Jiang, Chin Ken Wong, Martina H Stenzel
ABSTRACT
A novel gene transfection vector was fabricated based on the conjugation of human serum albumin (HSA) and maleimide end functionalized poly[(N,N-dimethylamino) ethyl methacrylate] (PDMAEMA). The bioconjugation was achieved in a site-specific manner to yield well-defined polymer-protein conjugates. The biohybrid was able to bind DNA with high affinity resulting in nanoparticles with a HSA shell. This paper mainly focuses on the influence of polymeric chain length on the particle properties and their drug-carrying ability to deliver oligonucleotides into breast cancer cells. The cytotoxic agent of interest, ISIS5132, is an oligonucleotide which disrupts DNA function within the cell. There was no evidence that the polymeric chain length had any effects on the conjugation efficiency and the subsequent condensation ability of the conjugates to oligonucleotide. However, the polymeric chain length had an obvious effect on the size of the complex micelles. Low molecular weights only led to loosely compacted complexes with the oligonucleotide, while large molecular weight led to well-defined nanoparticle structures. More importantly, it was found that the variation in the length of the PDMAEMA block resulted in a change in cytotoxicity of the drug loaded complex micelle. That is, the concentration of 50% inhibition (IC50 ) of the complex micelle on MDA-MB-231 and MCF-7 cells reached the lowest value at a chain length of around 21 000 g mol(-1) . The IC50 value increased when the polymeric chain length was shorter (8000 g mol(-1) and 10 000 g mol(-1) ) while it increased again when PDMAMEA of M¯n = 47 000 g mol(-1) , probably due to insufficient release of the drug. These result were reflected when investigating the performance of the polyplex using MCF-7 multicellular tumor spheroids, where again the medium PDMAEMA chain length led to the best delivery vehicle for ISIS5132.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Furan, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
Maleic anhydride, 99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS