Merck
  • Nanophotonic detection of freely interacting molecules on a single influenza virus.
Nanophotonic detection of freely interacting molecules on a single influenza virus.
Scientific reports (2015-07-15)
Pilgyu Kang, Perry Schein, Xavier Serey, Dakota O'Dell, David Erickson
ABSTRACT
Biomolecular interactions, such as antibody-antigen binding, are fundamental to many biological processes. At present, most techniques for analyzing these interactions require immobilizing one or both of the interacting molecules on an assay plate or a sensor surface. This is convenient experimentally but can constrain the natural binding affinity and capacity of the molecules, resulting in data that can deviate from the natural free-solution behavior. Here we demonstrate a label-free method for analyzing free-solution interactions between a single influenza virus and specific antibodies at the single particle level using near-field optical trapping and light-scattering techniques. We determine the number of specific antibodies binding to an optically trapped influenza virus by analyzing the change of the Brownian fluctuations of the virus. We develop an analytical model that determines the increased size of the virus resulting from antibodies binding to the virus membrane with uncertainty of ± 1-2 nm. We present stoichiometric results of 26 ± 4 (6.8 ± 1.1 attogram) anti-influenza antibodies binding to an H1N1 influenza virus. Our technique can be applied to a wide range of molecular interactions because the nanophotonic tweezer can handle molecules from tens to thousands of nanometers in diameter.
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Anti-Influenza A Antibody, H1N1 Antigen, clone 9B3.2, ascites fluid, clone 9B3.2, Chemicon®
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid