Merck
  • PS/CTAB/silica composites from room temperature polymerization of high internal phase emulsion gels.
PS/CTAB/silica composites from room temperature polymerization of high internal phase emulsion gels.
Journal of colloid and interface science (2015-04-22)
Tan Zhang, Gu Xu, Zhe-Fei Li, Oren Regev, Madhubhashini Maddumaarachchi, Frank D Blum
ABSTRACT
Polystyrene (PS)/cetyltrimethylammonium bromide (CTAB)/silica composites were prepared by CTAB-catalyzed in-situ polymerization of high internal phase emulsion gels (HIPE gels) at room temperature. The room temperature approach in HIPE gels yielded mono-dispersed PS/CTAB/silica spherical composite particles with sizes of 121 and 155nm. The complex microscopic internal structures of these particles were determined with the use of temperature-modulated differential scanning calorimetry (TMDSC). CTAB, in particular, was identified in different environments within the composite particles based on its different crystallization temperatures observed on cooling: a thin layer at the surface of the fumed silica (TC,CTAB=74°C), multilayers (TC,CTAB=82°C), and a mixed PS/CTAB shell (TC,CTAB=65°C). The amounts of each component were estimated from the enthalpies of the crystallization. The PS/CTAB mixed shell contained CTAB on the surface of the particle plus internal CTAB domains.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%