Merck
  • Growth and Optical Properties of High-Quality Monolayer WS2 on Graphite.
Growth and Optical Properties of High-Quality Monolayer WS2 on Graphite.
ACS nano (2015-03-27)
Yu Kobayashi, Shogo Sasaki, Shohei Mori, Hiroki Hibino, Zheng Liu, Kenji Watanabe, Takashi Taniguchi, Kazu Suenaga, Yutaka Maniwa, Yasumitsu Miyata
ABSTRACT
Atomic-layer transition metal dichalcogenides (TMDCs) have attracted appreciable interest due to their tunable band gap, spin-valley physics, and potential device applications. However, the quality of TMDC samples available still poses serious problems, such as inhomogeneous lattice strain, charge doping, and structural defects. Here, we report on the growth of high-quality, monolayer WS2 onto exfoliated graphite by high-temperature chemical vapor deposition (CVD). Monolayer-grown WS2 single crystals present a uniform, single excitonic photoluminescence peak with a Lorentzian profile and a very small full-width at half-maximum of 21 meV at room temperature and 8 meV at 79 K. Furthermore, in these samples, no additional peaks are observed for charged and/or bound excitons, even at low temperature. These optical responses are completely different from the results of previously reported TMDCs obtained by mechanical exfoliation and CVD. Our findings indicate that the combination of high-temperature CVD with a cleaved graphite surface is an ideal condition for the growth of high-quality TMDCs, and such samples will be essential for revealing intrinsic physical properties and for future applications.
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Pricing and availability is not currently available.