Merck
  • Characterization of five putative aspartate aminotransferase genes in the N2-fixing heterocystous cyanobacterium Anabaena sp. strain PCC 7120.
Characterization of five putative aspartate aminotransferase genes in the N2-fixing heterocystous cyanobacterium Anabaena sp. strain PCC 7120.
Microbiology (Reading, England) (2015-03-27)
Xinyi Xu, Liping Gu, Ping He, Ruanbao Zhou
ABSTRACT
Aspartate and glutamate are two key amino acids used in biosynthesis of many amino acids that play vital role in cellular metabolism. Aspartate aminotransferases (AspATs) are required for channelling nitrogen (N(2)) between Glu and Asp in all life forms. Biochemical and genetic characterization of AspATs have been lacking in N(2)-fixing cyanobacteria. In this report, five putative AspAT genes (alr1039, all2340, alr2765, all4327 and alr4853) were identified in the N(2)-fixing heterocystous cyanobacterium Anabaena sp. PCC 7120. Five recombinant C-terminal hexahistidine-tagged AspATs (AspAT-H(6)) were overexpressed in Escherichia coli and purified to homogeneity. Biochemical analysis demonstrated that these five putative AspATs have authentic AspAT activity in vitro using aspartate as an amino donor. However, the enzymic activities of the five AspATs differed in vitro. Alr4853-H(6) showed the highest AspAT activity, while the enzymic activity for the other four AspATs ranged from 6.5 to 53.7 % activity compared to Alr4853 (100 %). Genetic characterization of the five AspAT genes was also performed by inactivating each individual gene. All of the five AspAT knockout mutants exhibited reduced diazotrophic growth, and alr4853 was further identified to be a Fox gene (requiring fixed N(2) for growth in the presence of oxygen). Four out of five P(aspAT)-gfp transcriptional fusions were constitutively expressed in both diazotrophic and nitrate-dependent growth conditions. Quantitative reverse transcriptase PCR showed that alr4853 expression was increased by 2.3-fold after 24 h of N(2) deprivation. Taken together, these findings add to our understanding of the role of AspATs in N(2)-fixing within heterocystous cyanobacteria.
Sigma-Aldrich
AST Activity Assay Kit, sufficient for 100 colorimetric tests