Merck
  • MDC1 Enhances Estrogen Receptor-mediated Transactivation and Contributes to Breast Cancer Suppression.
MDC1 Enhances Estrogen Receptor-mediated Transactivation and Contributes to Breast Cancer Suppression.
International journal of biological sciences (2015-07-30)
Renlong Zou, Xinping Zhong, Chunyu Wang, Hongmiao Sun, Shengli Wang, Lin Lin, Shiying Sun, Changci Tong, Hao Luo, Peng Gao, Yanshu Li, Tingting Zhou, Da Li, Liu Cao, Yue Zhao
ABSTRACT
Estrogen receptor α (ERα) is a key transcriptional factor in the proliferation and differentiation in mammary epithelia and has been determined to be an important predictor of breast cancer prognosis and therapeutic target. Meanwhile, diverse transcriptional co-regulators of ERα play crucial and complicated roles in breast cancer progression. Mediator of DNA damage checkpoint 1 (MDC1) has been identified as a critical upstream mediator in the cellular response to DNA damage, however, some non-DNA damage responsive functions of MDC1 haven't been fully defined. In this study, we have identified MDC1 as a co-activator of ERα in breast cancer cells and demonstrated that MDC1 associates with ERα. MDC1 was also recruited to estrogen response element (ERE) of ERα target gene. Knockdown of MDC1 reduced the transcription of the endogenous ERα target genes, including p21. MDC1 depletion led to the promotion of breast cancer progression, and the expression of MDC1 is lower in breast cancer. Taken together, these results suggested that MDC1 was involved in the enhancement of ERα-mediated transactivation in breast cancer cells. This positive regulation by MDC1 might contribute to the suppression of breast cancer progression by acting as a barrier of positive to negative ERα function transformation.
Sigma-Aldrich
MISSION® esiRNA, targeting human ESR1
Sigma-Aldrich
MISSION® esiRNA, targeting human EREG
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ereg
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)