Merck
  • Immunolocalization of fibroblast growth factor receptors 1 and 2 in mouse palate development.
Immunolocalization of fibroblast growth factor receptors 1 and 2 in mouse palate development.
Plastic and reconstructive surgery (2001-06-08)
S Lee, C A Crisera, S Erfani, T S Maldonado, J J Lee, S L Alkasab, M T Longaker
ABSTRACT
Recent evidence has implicated mutations of fibroblast growth factor receptors (FGF-R) in the pathogenesis of craniosynostotic syndromes. Cleft palate can be a component of such syndromes. The expression of FGF-R1 and FGF-R2 has been delineated in normally developing cranium, where they seem to regulate cellular differentiation and proliferation, respectively. The specific role of fibroblast growth factor signaling in mammalian palate development is unclear. The authors investigated the patterns of expression of FGF-R1 and FGF-R2 throughout mouse palatal development in the embryo. Time-dated CD-1 mouse heads (n = 135) were harvested at embryonic ages 12.5, 13.5, 14.5, 15.5, and 16.5 days (term gestation = 19.5 days), fixed in paraformaldehyde, embedded in paraffin, and sectioned. In addition, paired palatal shelves (n = 30) were isolated by means of microdissection from embryonic day--13.5 embryos, grown on Millipore filters in serum-free medium in vitro for 24, 48, 72, or 96 hours and processed for histological analysis. Immunohistochemical analysis for FGF-R1 and FGF-R2 was performed on the in vivo and in vitro specimens. FGF-R1 and FGF-R2 were found to be specifically expressed in the epithelium of the developing palatal shelves from the time of their outgrowth from the maxillary processes through completion of fusion in vivo and in vitro. Expression of both receptors was particularly strong during the phases of medial epithelial-medial epithelial contact between the individual shelves, through the formation of the medial epithelial seam, to the ultimate dissolution of the seam. Such a pattern of expression seems to implicate fibroblast growth factor signaling in the regulation of the critical phase of fusion of the bilateral shelves. The expression of both FGF-R1 and FGF-R2 in the lateral palatal mesenchyme, where such secondary structures as tooth primordia and bone begin to appear, also suggests a role for fibroblast growth factor signaling in the induction of ongoing differentiation and maturation of the palate after fusion. These data suggest that fibroblast growth factor signaling may play a role in the epithelial-mesenchymal interactions that dictate fusion and maturation of the developing palate. Furthermore, the data are consistent with the correlation of cleft palate formation with aberrant fibroblast growth factor signaling.