Merck
  • An efficient synthesis of achiral and chiral 1,2,4-triazolium salts: bench stable precursors for N-heterocyclic carbenes.
An efficient synthesis of achiral and chiral 1,2,4-triazolium salts: bench stable precursors for N-heterocyclic carbenes.
The Journal of organic chemistry (2005-07-02)
Mark S Kerr, Javier Read de Alaniz, Tomislav Rovis
ABSTRACT
[reaction: see text] The promising utility of triazolyl N-heterocyclic carbene catalysts in umpolung aldehyde chemistry requires a straightforward reliable synthesis from readily available materials. Herein, we describe the synthesis of a variety of triazolyl N-heterocyclic carbene precursors. The reactions commence from commercially available amino acids and proceed in 44-68% overall yields. The N-heterocyclic salts are air-stable crystalline solids that can be stored with no special precaution and can generate the active catalyst when treated with an appropriate base.
Sigma-Aldrich
Triethyl orthoformate, anhydrous, 98%