Merck
  • Cloning and expression of an inducible lymphoid-specific, protein tyrosine phosphatase (HePTPase).
Cloning and expression of an inducible lymphoid-specific, protein tyrosine phosphatase (HePTPase).
European journal of immunology (1992-01-01)
B Zanke, H Suzuki, K Kishihara, L Mizzen, M Minden, A Pawson, T W Mak
ABSTRACT
Tyrosine phosphorylation and dephosphorylating events have been shown to be central to the process of growth regulation and signal transduction. We report here, the identification of a new gene with a tyrosine phosphatase domain (EC 3.1.3.48) which is expressed exclusively in thymus and spleen. A cDNA of 2760 bp encodes a 339-amino acid, intracellular, single-domain tyrosine phosphatase. When expressed as a glutathionine-S-transferase fusion protein, efficient lysis of p-nitrophenyl phosphate is noted, indicating in vitro enzymatic activity of the cloned gene product. Normal mouse lymphocytes increase mRNA expression 10-15-fold upon stimulation with phytohemagglutinin, concanavalin A, lipopolysaccharide or anti-CD3 monoclonal antibody. This new hematopoietic tyrosine phosphatase, (HePTP), may play a role in the regulation of T and B lymphocyte development and signal transduction.