Merck
  • Synthesis, structure and biological activities of cobalt(II) and zinc(II) coordination compounds with 2-benzimidazole derivatives.
Synthesis, structure and biological activities of cobalt(II) and zinc(II) coordination compounds with 2-benzimidazole derivatives.
Journal of inorganic biochemistry (2008-02-29)
Horacio López-Sandoval, Milton E Londoño-Lemos, Raúl Garza-Velasco, Israel Poblano-Meléndez, Pilar Granada-Macías, Isabel Gracia-Mora, Norah Barba-Behrens
ABSTRACT
In this work we present the synthesis, structural and spectroscopic characterisation of a series of cobalt(II) and zinc(II) coordination compounds with benzimidazole (bz) and its 2-benzimidazole derivatives: 2-aminobenzimidazole (2ab), albendazole (abz) and tris(2-benzimidazolylmethyl)amine (ntb). The compounds were evaluated for their in vitro antimicrobial activity against Staphylococcus aureus, Micrococcus luteus, Salmonella typhi, Pseudomonas aeruginosa, Escherichia coli and Proteus vulgaris. Their cytotoxic activity was also evaluated using human cancer lines, HeLa, HCT-15 and SKLU-1. The halide tetrahedral compounds [Co(bz)2Br2] 3, [Zn(2ab)2Cl2].0.5H2O 11, [Co(abz)Cl2(H2O)].3H2O 14, [Co(abz)Br2(H2O)] 15, [Zn(abz)Cl2(H2O)].3H2O 17 and [Zn(abz)Br2(H2O)].H2O 18 displayed similar minimal inhibition concentration (MIC) values against Micrococcus luteus and Escherichia coli, comparable to those of amoxicillin and chloramphenicol. Additionally, 11 showed a wide range of activity towards Gram(+) and Gram(-) microorganisms. The tetradentate ntb and its trigonal bipyramidal cobalt(II) and zinc(II) compounds were active, regardless of the anion present in the complex. Compound [Co(abz)Cl2(H2O)].3H2O 14 showed promising activity in HeLa cells, while [Co(ntb)Br]Br.H2O 21 inhibited Hela and HCT-15 cell lines.
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, reagent grade, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, ACS reagent, ≥98%
Pricing and availability is not currently available.