Merck
  • Preparation, characterization, and preliminary antibrowning evaluations of norartocarpetin microemulsions.
Preparation, characterization, and preliminary antibrowning evaluations of norartocarpetin microemulsions.
Journal of agricultural and food chemistry (2015-01-21)
Zong-Ping Zheng, Xue Dong, Kun Yuan, Shan Lan, Qin Zhu, Mingfu Wang, Jie Chen
ABSTRACT
Norartocarpetin is a flavone widely distributed in Moraceae plants with strong tyrosinase inhibitory activity. However, its poor solubility in aqueous systems and in food grade solvents (oils) limits its extensive applications. The aim of this study was to improve the solubility of norartocarpetin by developing an oil-in-water (o/w) microemulsion with food grade components. A microemulsion was developed and characterized, and its chemical and physical stabilities were also evaluated within 8 weeks. Using the construction of pseudoternary phase diagrams, the optimized formulation of 2% w/w of ethyl oleate, 12% w/w of Tween 80, 6% w/w of polyethylene glycol 400, and 80% w/w of water was obtained, with a maximum solubility of norartocarpetin up to 1.73 ± 0.21 mg/mL, at least a 1000-fold increase in solubility. The microemulsion base and norartocarpetin-loaded microemulsion were demonstrated to be stable after accelerated and long-term conditions (8 weeks). The norartocarpetin microemulsion still showed strong tyrosinase inhibitory activity and antibrowning effect on fresh-cut apple slices. These combined results indicated that norartocarpetin microemulsion may be suitable as an antibrowning agent for fresh-cut fruits.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ethyl oleate, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Ethyl oleate, technical grade, 70%
Sigma-Aldrich
Ethyl oleate, tested according to Ph. Eur.
Sigma-Aldrich
4-Hexylresorcinol, 98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Ethyl oleate, natural, ≥85%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
L-Ascorbic acid, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material