Merck
  • A hemicellulose-bound form of silicon with potential to improve the mechanical properties and regeneration of the cell wall of rice.
A hemicellulose-bound form of silicon with potential to improve the mechanical properties and regeneration of the cell wall of rice.
The New phytologist (2015-01-24)
Congwu He, Jie Ma, Lijun Wang
ABSTRACT
Silicon (Si) plays a large number of diverse roles in plants, but the structural and chemical mechanisms operating at the single-cell level remain unclear. We isolate the cell walls from suspension-cultured individual cells of rice (Oryza sativa) and fractionate them into three main fractions including cellulose (C), hemicellulose (HC) and pectin (P). We find that most of the Si is in HC as determined by inductively coupled plasma-mass spectrometry (ICP-MS), where Si may covalently crosslink the HC polysacchrides confirmed by X-ray photoelectron spectroscopy (XPS). The HC-bound form of Si could improve both the mechanical property and regeneration of the cell walls investigated by a combination of atomic force microscopy (AFM) and confocal laser scanning microscopy (CLSM). This study provides further evidence that HC could be the major ligand bound to Si, which broadens our understanding of the chemical nature of 'anomalous' Si in plant cell walls.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%