Merck
  • A novel compound, NK150460, exhibits selective antitumor activity against breast cancer cell lines through activation of aryl hydrocarbon receptor.
A novel compound, NK150460, exhibits selective antitumor activity against breast cancer cell lines through activation of aryl hydrocarbon receptor.
Molecular cancer therapeutics (2014-12-20)
Kazuteru Fukasawa, Shigehide Kagaya, Sakiko Maruyama, Shunsuke Kuroiwa, Kuniko Masuda, Yoshio Kameyama, Yoshitaka Satoh, Yuichi Akatsu, Arihiro Tomura, Kiyohiro Nishikawa, Shigeo Horie, Yuh-ichiro Ichikawa
ABSTRACT
Antiestrogen agents are commonly used to treat patients with estrogen receptor (ER)-positive breast cancer. Tamoxifen has been the mainstay of endocrine treatment for patients with early and advanced breast cancer for many years. Following tamoxifen treatment failure, however, there are still limited options for subsequent hormonal therapy. We discovered a novel compound, NK150460, that inhibits 17β-estradiol (E2)-dependent transcription without affecting binding of E2 to ER. Against our expectations, NK150460 inhibited growth of not only most ER-positive, but also some ER-negative breast cancer cell lines, while never inhibiting growth of non-breast cancer cell lines. Cell-based screening using a random shRNA library, identified aryl hydrocarbon receptor nuclear translocator (ARNT) as a key gene involved in NK150460's antitumor mechanism. siRNAs against not only ARNT but also its counterpart aryl hydrocarbon receptor (AhR) and their target protein, CYP1A1, dramatically abrogated NK150460's growth-inhibitory activity. This suggests that the molecular cascade of AhR/ARNT plays an essential role in NK150460's antitumor mechanism. Expression of ERα was decreased by NK150460 treatment, and this was inhibited by an AhR antagonist. Unlike two other AhR agonists now undergoing clinical developmental stage, NK150460 did not induce histone H2AX phosphorylation or p53 expression, suggesting that it did not induce a DNA damage response in treated cells. Cell lines expressing epithelial markers were more sensitive to NK150460 than mesenchymal marker-expressing cells. These data indicate that NK150460 is a novel AhR agonist with selective antitumor activity against breast cancer cell lines, and its features differ from those of the other two AhR agonists.
Product Description
Supelco
3-Methylcholanthrene solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
β-Naphthoflavone, ≥98%
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Supelco
3-Methylcholanthrene, analytical standard
Sigma-Aldrich
Leu-Leu
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-Methylcholanthrene, 98%