Merck
  • Reconstruction of hepatic stellate cell-incorporated liver capillary structures in small hepatocyte tri-culture using microporous membranes.
Reconstruction of hepatic stellate cell-incorporated liver capillary structures in small hepatocyte tri-culture using microporous membranes.
Journal of tissue engineering and regenerative medicine (2012-10-23)
Junichi Kasuya, Ryo Sudo, Genta Masuda, Toshihiro Mitaka, Mariko Ikeda, Kazuo Tanishita
ABSTRACT
In liver sinusoids, hepatic stellate cells (HSCs) locate the outer surface of microvessels to form a functional unit with endothelia and hepatocytes. To reconstruct functional liver tissue in vitro, formation of the HSC-incorporated sinusoidal structure is essential. We previously demonstrated capillary formation of endothelial cells (ECs) in tri-culture, where a polyethylene terephthalate (PET) microporous membrane was intercalated between the ECs and hepatic organoids composed of small hepatocytes (SHs), i.e. hepatic progenitor cells, and HSCs. However, the high thickness and low porosity of the membranes limited heterotypic cell-cell interactions, which are essential to form HSC-EC hybrid structures. Here, we focused on the effective use of the thin and highly porous poly( d, l-lactide-co-glycolide) (PLGA) microporous membranes in SH-HSC-EC tri-culture to reconstruct the HSC-incorporated liver capillary structures in vitro. First, the formation of EC capillary-like structures was induced on Matrigel-coated PLGA microporous membranes. Next, the membranes were stacked on hepatic organoids composed of small SHs and HSCs. When the pore size and porosity of the membranes were optimized, HSCs selectively migrated to the EC capillary-like structures. This process was mediated in part by platelet-derived growth factor (PDGF) signalling. In addition, the HSCs were located along the outer surface of the EC capillary-like structures with their long cytoplasmic processes. In the HSC-incorporated capillary tissues, SHs acquired high levels of differentiated functions, compared to those without ECs. This model will provide a basis for the construction of functional, thick, vascularized liver tissues in vitro.
Product Description
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
SAFC
HEPES
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetic acid, analytical standard
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
SAFC
L-Proline
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
SAFC
HEPES
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer