Merck
  • Oleanane triterpenes as protein tyrosine phosphatase 1B (PTP1B) inhibitors from Camellia japonica.
Oleanane triterpenes as protein tyrosine phosphatase 1B (PTP1B) inhibitors from Camellia japonica.
Phytochemistry (2014-05-13)
Mohammad Nasir Uddin, Govinda Sharma, Jun-Li Yang, Hong Seok Choi, Seong-Il Lim, Keon Wook Kang, Won Keun Oh
ABSTRACT
Protein tyrosine phosphatase 1B (PTP1B) plays a key role in metabolic signaling, thereby making it an exciting drug target for type 2 diabetes and obesity. Besides, there is substantial evidence that shows its overexpression is involved in breast cancer, which suggests that selective PTP1B inhibition might be effective in breast cancer treatment. As part of our continuous research on PTP1B inhibitors from medicinal plants, four oleanane-type triterpenes were isolated from an EtOAc-soluble extract of fruit peels of Camellia japonica (Theaceae), together with 6 previously known compounds of this class. Their structures were determined on the basis of spectroscopic data analysis (UV, IR, (1)H and (13)CNMR, HMBC, HSQC, NOESY, and MS). All isolates were evaluated for their inhibitory effects on PTP1B, as well as their cytotoxic effects against human breast cancer cell lines MCF7, MCF7/ADR, and MDA-MB-231. Several compounds with OH-3 or/and COOH-28 functionalities showed strong PTP1B inhibitory activity (IC50 values ranging from 3.77±0.11 to 6.40±0.81 μM) as well as significant cytotoxicity (IC50 values ranging from 0.51±0.05 to 13.55±1.44 μM).
Sigma-Aldrich
PTP1B (1-321)/PTPN1 Active human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)