Merck
  • Characterization of human lymphoblastoid cell lines as a novel in vitro test system to predict the immunotoxicity of xenobiotics.
Characterization of human lymphoblastoid cell lines as a novel in vitro test system to predict the immunotoxicity of xenobiotics.
Toxicology letters (2014-12-30)
Tijana Markovič, Martina Gobec, David Gurwitz, Irena Mlinarič-Raščan
ABSTRACT
Evaluating immunomodulatory effects of xenobiotics is an important component of the toxicity studies. Herein we report on the establishment of a novel invitro test system for the immunotoxicity screening of xenobiotics based on human lymphoblastoid cell lines (LCLs). Four immunotoxic compounds; tributyltin chloride, cyclosporine A, benzo(a)pyrene and verapamil hydrochloride, as well as three immune-inert compounds; urethane, furosemide and mannitol were selected for characterization. The treatment of LCLs with immunosuppressive compounds resulted in reduced viability. The IC50 values determined in human LCLs were in agreement with the data obtained for human peripheral mononuclear cells. Since cytokine production reflects lymphocytes responses to external stimuli, we evaluated the functional responses of LCLs by monitoring their pro-inflammatory and immunoregulatory cytokine production. Our findings prove that LCLs allowed for reliable differentiation between immunomodulatory and immune-inert compounds. Hence, pre-treatment with immunomodulatory compounds led to a decrease in the production of pro-inflammatory TNFα, IL-6 and immunoregulatory IL-2, IL-4, IL-10 and IFNγ cytokines, when compared to untreated ionomycin/PMA stimulated cells. Moreover, testing a panel of ten LCLs derived from unrelated healthy individuals reflects inter-individual variability in response to immunomodulatory xenobiotics. In conclusion, LCLs provide a novel alternative method for the testing of the immunotoxic effects of xenobiotics.
Product Description
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Millipore
D-Mannitol, ACS reagent, ≥99.0%, suitable for microbiology
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Furosemide, European Pharmacopoeia (EP) Reference Standard
USP
Furosemide, United States Pharmacopeia (USP) Reference Standard
Supelco
Urethane, analytical standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Tributyltin chloride, 96%