Merck
  • Mitochondrial division inhibitor-1 induces mitochondrial hyperfusion and sensitizes human cancer cells to TRAIL-induced apoptosis.
Mitochondrial division inhibitor-1 induces mitochondrial hyperfusion and sensitizes human cancer cells to TRAIL-induced apoptosis.
International journal of oncology (2014-09-02)
Mamoru Akita, Miki Suzuki-Karasaki, Kyoko Fujiwara, Chinatsu Nakagawa, Masayoshi Soma, Yukihiro Yoshida, Toyoko Ochiai, Yasuaki Tokuhashi, Yoshihiro Suzuki-Karasaki
ABSTRACT
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a promising candidate for cancer treatment, but some cancer cell types are resistant to TRAIL cytotoxicity. Therefore, overcoming this resistance is necessary for effective TRAIL therapy. Mitochondrial morphology is important for the maintenance of cell function and survival, and is regulated by the delicate balance between fission and fusion. However, the role of mitochondrial morphology dynamics in TRAIL-induced apoptosis is unknown. Here we show that mitochondrial division inhibitor-1 (mdivi-1), an inhibitor of dynamin-related protein1 (Drp1), modulates mitochondrial morphology and TRAIL-induced apoptosis in human cancer cells. mdivi-1 treatment (≥12.5 µM) caused dose- and time‑dependent cell death in malignant melanoma, lung cancer and osteosarcoma cells, while sparing normal cells. mdivi-1 also sensitized cancer cells to TRAIL-induced apoptosis. This potentiation of apoptosis occurred through a caspase-depependent mechanism including the mitochondrial and endoplasmic reticulum (ER) stress pathways. Mdivi-1 potentiated mitochondrial oxidative stress, a major cause of mitochondrial and ER stresses, as evidenced by increases in mitochondrial reactive oxygen species levels, mitochondrial mass, and cardiolipin oxidation. Live cell fluorescence imaging using MitoTracker Red CMXRos revealed that Mdivi-1 caused substantial mitochondrial hyperfusion. Moreover, silencing of Drp1 expression also caused mitochondrial hyperfusion and sensitized cancer cells to TRAIL-induced apoptosis. Our results suggest that cancer cells are more vulnerable than normal cells to a perturbation in mitochondrial morphology dynamics and that this higher susceptibility can be exploited to selectively kill cancer cells and sensitize to TRAIL.
Sigma-Aldrich
TRAIL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE and HPLC), lyophilized powder
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFRSF10A
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
TRAIL human, recombinant, expressed in NSO cells, >97% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Supelco
Rotenone, PESTANAL®, analytical standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Human TNFRSF10A ELISA Kit